Learn More
3M™ Versaflo™ M-300 Series PAPR Accessory, Forehead Seal
Product Code. 15740078 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-300 Series Hardhats and M-400 Series Helmets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15740078 M-300 Series Hardhats and M-400 Series Helmets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15740078 Supplier 3M™ Supplier No. 7100063941
  • ONLINE EXCLUSIVE PRICE
    62.00€
    42.17€ / pezzi 1
    Risparmia 19.83€
    31% Sconto
Spedizione stimata: 07-01-2027
Forehead seal is a replacement part for Versaflo™ M-300 Series hardhats.
  • 3M Catalog# M-354
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Disposable Reusable
Product Type Forehead Seal
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) M-300 Series Hardhats and M-400 Series Helmets
Model M-354