Learn More
3M™ Versaflo™ Supplied Air Regulator Assembly, V-500E
Codice prodotto. 31605235 Sfoglia Tutto 3M Prodotti
Cambia vista
Click to view available options
Product Type:
Adjustable Regulator
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
31605235 Adjustable Regulator Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31605235 Fornitore 3M™ N. del fornitore 7100342003
  • ONLINE EXCLUSIVE PRICE
    820.00€
    524.67€ / pezzi 1
    Risparmia 295.33€
    36% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 09-12-2026
V-500E Series supplied air regulator assembly provides reliable respiratory protection with adjustable airflow and continuous compressed air for comfort and durability in tough environments.
  • 3M™ Versaflo™ Supplied Air Respirator System V-500E Series provides reliable respiratory protection and comfort for workers across various industrial settings.
  • Delivers a continuous flow of air from a compressed air source, ensuring a steady and consistent air supply.
  • Compatible with a wide range of 3M headtops, including hoods and helmets, for flexible and versatile use.
  • Features adjustable airflow control, allowing users to customize airflow according to their specific needs and comfort preferences.
  • Offers effective protection against airborne contaminants for enhanced workplace safety.
  • Suitable for industries such as manufacturing, construction, and chemical processing.
TRUSTED_SUSTAINABILITY
Specifica
Materiale Plastic
Product Type Adjustable Regulator