Learn More
3M™ Versaflo™ Supplied Air Regulator Paint Hood Kit, V-500E-PHK
Product Code. 31605230 Shop All 3M Products
Change view
Click to view available options
Product Type:
Supplied Air Regulator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31605230 Supplied Air Regulator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31605230 Supplier 3M™ Supplier No. 7100342201
  • ONLINE EXCLUSIVE PRICE
    1430.00€
    914.98€ / pezzi 1
    Risparmia 515.02€
    36% Sconto
 Restricted Item
Spedizione stimata: 02-12-2026
V-500E Series supplied air regulator assembly provides reliable respiratory protection with adjustable airflow and continuous compressed air for comfort and durability in tough environments.
  • 3M™ Versaflo™ Supplied Air Respirator System V-500E Series provides dependable respiratory protection and comfort for workers in diverse industrial settings.
  • Delivers a continuous flow of air from a compressed air source, ensuring a steady and reliable air supply.
  • Compatible with a wide range of 3M headtops, including hoods and helmets, for versatile application.
  • Features adjustable airflow control, allowing users to customize airflow based on individual needs and preferences.
  • Protects against airborne contaminants to enhance safety and performance.
  • Ideal for use in industries such as manufacturing, construction, and chemical processing.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Supplied Air Regulator