Learn More
4-(1-Adamantyl)phenol, TRC
Product Code. 30317488
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317488 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30317488 Supplier Toronto Research Chemicals Supplier No. TRCA2200001G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(1-Adamantyl)phenol
CAS 29799-07-3
Molecular Formula C16H20O
Quantity 1 g
Synonym p-1-Adamantyl-phenol, 1-(4-Hydroxyphenyl)adamantane, 1-(p-Hydroxyphenyl)adamantane, 4-(Hydroxyphenyl)adamantane, NSC 111653, p-1-Adamantylphenol, 4-Tricyclo[3.3.1.13,7]dec-1-yl-phenol
InChI Formula InChI=1S/C16H20O/c17-15-3-1-14(2-4-15)16-8-11-5-12(9-16)7-13(6-11)10-16/h1-4,11-13,17H,5-10H2
SMILES OC(=O)[C@@H](N)Cc1ncnc1
IUPAC Name 4-(1-adamantyl)phenol
Molecular Weight (g/mol) 228.33
Formula Weight 228.1514