missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(1-Piperidinyl)-4-thiazolyl]benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number p-7039384
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
C15H19BN2O2
MFCD22682994
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
C15H19BN2O2
MFCD22682994

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-[2-(1-Piperidinyl)-4-thiazolyl]benzeneboronic acid
Quantità 1 g
Peso formulazione 270.14
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'