Learn More
4-(2-Phthalimidoethoxy)acetoacetic Acid Ethyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-(2-Phthalimidoethoxy)acetoacetic Acid Ethyl Ester |
| CAS | 88150-75-8 |
| Formula molecolare | C16H17NO6 |
| Quantità | 250 mg |
| Sinonimo | Ethyl 4-(2-Phthalimidoethoxy)acetoacetate, 4-[2-(1,3-Dioxo-1,3-dihydroisoindol-2-yl)ethoxy]-3-oxobutanoic Acid Ethyl Ester, 4-[2-(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]-3-oxobutanoic Acid Ethyl Ester, Ethyl 4-[2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]-3-oxobutanoate (ACI), 4-(2-Phthalimidoethoxy)acetoacetic acid ethyl ester |
| InChI Formula | InChI=1S/C16H17NO6/c1-2-23-14(19)9-11(18)10-22-8-7-17-15(20)12-5-3-4-6-13(12)16(17)21/h3-6H,2,7-10H2,1H3 |
| SMILES | O=C(OCC)CC(=O)COCCN1C(=O)C=2C=CC=CC2C1=O |
| IUPAC Name | Ethyl 4-[2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]-3-oxobutanoate |
| Molecular Weight (g/mol) | 319.31 |
| Peso formulazione | 319.11 |