missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7046942
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
338-03-4
C5H7F3O3
172.103
MFCD00039531
JCJGOWYCFRASTM-UHFFFAOYSA-N
3-trifluoromethyl-3-hydroxybutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methylbutyric acid, 3-hydroxy-3-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methyl-butyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-methyl, pubchem12644, acmc-20an2r, 3-hydroxy-3-trifluoromethylbutyric acid, 3-hydroxy-3-trifluoromethyl butyric acid
136168
4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid
CC(CC(=O)O)(C(F)(F)F)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
338-03-4
C5H7F3O3
172.103
MFCD00039531
JCJGOWYCFRASTM-UHFFFAOYSA-N
3-trifluoromethyl-3-hydroxybutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methylbutyric acid, 3-hydroxy-3-methyl-4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-3-hydroxy-3-methyl-butyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-methyl, pubchem12644, acmc-20an2r, 3-hydroxy-3-trifluoromethylbutyric acid, 3-hydroxy-3-trifluoromethyl butyric acid
136168
4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid
CC(CC(=O)O)(C(F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid
Punto di fusione 39°C to 41°C
Punti di ebollizione 87°C to 90°C (0.5 mmHg)
Indice di rifrazione 1.388
Quantità 1 g
Numero UN UN3261
Peso formulazione 172.1
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'