missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Cyanophenyl)-2-cyclopropylthiazole, 97%, Thermo Scientific™

Catalog Number p-7039413
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1090552-63-8
C13H10N2S
226.297
MFCD11831275
SOAYBZVJEBLFEA-UHFFFAOYSA-N
4-2-cyclopropyl-1,3-thiazol-4-yl benzonitrile, 4-4-cyanophenyl-2-cyclopropylthiazole
35618808
4-(2-cyclopropyl-1,3-thiazol-4-yl)benzonitrile
C1CC1C2=NC(=CS2)C3=CC=C(C=C3)C#N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1090552-63-8
C13H10N2S
226.297
MFCD11831275
SOAYBZVJEBLFEA-UHFFFAOYSA-N
4-2-cyclopropyl-1,3-thiazol-4-yl benzonitrile, 4-4-cyanophenyl-2-cyclopropylthiazole
35618808
4-(2-cyclopropyl-1,3-thiazol-4-yl)benzonitrile
C1CC1C2=NC(=CS2)C3=CC=C(C=C3)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-(4-Cyanophenyl)-2-cyclopropylthiazole
Quantità 1 g
Numero UN UN3439
Peso formulazione 302.39
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'