missing translation for 'onlineSavingsMsg'
Learn More

4,4-Diethoxy-N,N-diethyl-1-butanamine, TRC

Product Code. 30351518
Change view
Click to view available options
Quantità:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30351518 250 mg 250mg
30349220 25 mg 25mg
30350483 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351518 Supplier Toronto Research Chemicals Supplier No. TRCD168970250MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97028-91-6
Formula molecolare C12H27NO2
Molecular Weight (g/mol) 217.35
Sinonimo 4-(Diethylamino)butanal Diethyl Acetal
IUPAC Name 4,4-diethoxy-N,N-diethylbutan-1-amine
SMILES CCOC(CCCN(CC)CC)OCC

Specifications

Materiale o nome chimico 4,4-Diethoxy-N,N-diethyl-1-butanamine
Quantità 250 mg
InChI Formula InChI=1S/C12H27NO2/c1-5-13(6-2)11-9-10-12(14-7-3)15-8-4/h12H,5-11H2,1-4H3
Peso formulazione 217.2042
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.