missing translation for 'onlineSavingsMsg'
Learn More

4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide, 97%, Thermo Scientific™

Catalog Number 15446036
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
196869-06-4
C15H15ClN2O4S
354.805
MFCD00026154
JVUHNUSVIURRRK-UHFFFAOYSA-N
n-4-5-chloro-2-methoxyphenyl sulfamoyl phenyl acetamide, n-4-5-chloro-2-methoxyphenyl amino sulfonyl phenyl acetamide, cbmicro_023955, cambridge id 5536192, 4'-5-chloro-2-methoxyphenylsulfamoyl acetanilide, 4'-n-5-chloro-2-methoxyphenyl sulfamoyl acetanilide
989525
N-[4-[(5-chloro-2-methoxyphenyl)sulfamoyl]phenyl]acetamide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)Cl)OC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
196869-06-4
C15H15ClN2O4S
354.805
MFCD00026154
JVUHNUSVIURRRK-UHFFFAOYSA-N
n-4-5-chloro-2-methoxyphenyl sulfamoyl phenyl acetamide, n-4-5-chloro-2-methoxyphenyl amino sulfonyl phenyl acetamide, cbmicro_023955, cambridge id 5536192, 4'-5-chloro-2-methoxyphenylsulfamoyl acetanilide, 4'-n-5-chloro-2-methoxyphenyl sulfamoyl acetanilide
989525
N-[4-[(5-chloro-2-methoxyphenyl)sulfamoyl]phenyl]acetamide
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC2=C(C=CC(=C2)Cl)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4'-(5-Chloro-2-methoxyphenylsulfamoyl)acetanilide
Quantità 1 g
Peso formulazione 354.81
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'