Learn More
4-Acetylbenzenesulfonyl Chloride, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-Acetylbenzenesulfonyl Chloride |
| CAS | 1788-10-9 |
| Formula molecolare | C8H7ClO3S |
| Quantità | 1 g |
| Sinonimo | p-Acetyl-benzenesulfonyl Chloride, 4-Acetylbenzene-1-sulfonyl Chloride, 4-Acetylbenzensulfonyl Chloride, 4-Acetylphenylsulfonyl Chloride, NSC 104128 |
| InChI Formula | InChI=1S/C8H7ClO3S/c1-6(10)7-2-4-8(5-3-7)13(9,11)12/h2-5H,1H3 |
| SMILES | CC(=O)c1ccc(cc1)S(=O)(=O)Cl |
| IUPAC Name | 4-acetylbenzenesulfonyl chloride |
| Molecular Weight (g/mol) | 218.66 |