Learn More
4-Anilino-4-oxobutanoic Acid, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-Anilino-4-oxobutanoic Acid |
| CAS | 102-14-7 |
| Formula molecolare | C10 H11 N O3 |
| Quantità | 500 mg |
| Sinonimo | Butanoic acid, 4-oxo-4-(phenylamino)-, Succinanilic acid (6CI,7CI,8CI), 4-Oxo-4-(phenylamino)butanoic acid, 4-Anilino-4-oxobutanoic acid, 4-Oxo-4-(N-phenylamino)butanoic acid, N-Phenylbutanedioic acid monoamide, N-Phenylsuccinamic acid, NSC 56471, Succinic acid monoanilide, Succinic monoanilide |
| InChI Formula | InChI=1S/C10H11NO3/c12-9(6-7-10(13)14)11-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14) |
| SMILES | OC(=O)CCC(=O)Nc1ccccc1 |
| IUPAC Name | 4-anilino-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 193.2 |
| Peso formulazione | 193.0739 |