missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-1-bromo-2-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7030526
43.33 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
185346-79-6
C13H10BrFO
281.124
MFCD09801041
BYTJTXKQBSNGCL-UHFFFAOYSA-N
4-benzyloxy-1-bromo-2-fluorobenzene, 4-benzyloxy-2-fluorobromobenzene, benzene,1-bromo-2-fluoro-4-phenylmethoxy, acmc-1c1vq, benzyloxy-4-bromo-3-fluorobenzene, 1-benzyloxy-4-bromo-3-fluorobenzene, benzyl 4-bromo-3-fluorophenyl ether, 1-bromo-2-fluoro-4-benzyloxy benzene
22273933
1-bromo-2-fluoro-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC(=C(C=C2)Br)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
185346-79-6
C13H10BrFO
281.124
MFCD09801041
BYTJTXKQBSNGCL-UHFFFAOYSA-N
4-benzyloxy-1-bromo-2-fluorobenzene, 4-benzyloxy-2-fluorobromobenzene, benzene,1-bromo-2-fluoro-4-phenylmethoxy, acmc-1c1vq, benzyloxy-4-bromo-3-fluorobenzene, 1-benzyloxy-4-bromo-3-fluorobenzene, benzyl 4-bromo-3-fluorophenyl ether, 1-bromo-2-fluoro-4-benzyloxy benzene
22273933
1-bromo-2-fluoro-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC(=C(C=C2)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Benzyloxy-1-bromo-2-fluorobenzene
Punto di fusione 33°C to 35°C
Quantità 5 g
Peso formulazione 281.13
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'