missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7037135
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
175357-73-0
C14H14BrNO
292.176
MFCD00017822
LJKLYFDMLGYRCA-UHFFFAOYSA-N
4-bromo-n-4-methoxyphenyl methyl aniline, 4-bromo-n-4-methoxybenzyl aniline, n-4-methoxybenzyl-4-bromoaniline, 4-bromo-phenyl-4-methoxy-benzyl-amine, 4-bromophenyl 4-methoxyphenyl methyl amine
775227
4-bromo-N-[(4-methoxyphenyl)methyl]aniline
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)Br
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
175357-73-0
C14H14BrNO
292.176
MFCD00017822
LJKLYFDMLGYRCA-UHFFFAOYSA-N
4-bromo-n-4-methoxyphenyl methyl aniline, 4-bromo-n-4-methoxybenzyl aniline, n-4-methoxybenzyl-4-bromoaniline, 4-bromo-phenyl-4-methoxy-benzyl-amine, 4-bromophenyl 4-methoxyphenyl methyl amine
775227
4-bromo-N-[(4-methoxyphenyl)methyl]aniline
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Bromo-N-(4-methoxybenzyl)aniline
Quantità 250 mg
Peso formulazione 292.18
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'