Learn More
4-(Bromomethyl)-α-methyl-benzeneacetic Acid Methyl Ester, TRC
Codice prodotto. 30337190
Cambia vista
Click to view available options
Quantità:
500 mg
Dimensione della confezione:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30337190 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30337190 Fornitore Toronto Research Chemicals N. del fornitore TRCB890440500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 4-(Bromomethyl)-alpha-methyl-benzeneacetic Acid Methyl Ester
CAS 99807-54-2
Formula molecolare C11 H13 Br O2
Quantità 500 mg
Sinonimo Loxoprofen Impurity 9
InChI Formula InChI=1S/C11H13BrO2/c1-8(11(13)14-2)10-5-3-9(7-12)4-6-10/h3-6,8H,7H2,1-2H3
SMILES COC(=O)C(C)c1ccc(CBr)cc1
IUPAC Name methyl 2-[4-(bromomethyl)phenyl]propanoate
Molecular Weight (g/mol) 257.124
Peso formulazione 256.01