Learn More
4'-Chloro-2'-fluoro-3'-methoxyacetophenone, 97%, Thermo Scientific™
Product Code. 15468925
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468925 250 mg 250mg
15487357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15468925 Supplier Thermo Scientific Alfa Aesar Supplier No. H34416.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1323966-44-4
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.61
MDL Number MFCD19687210
InChI Key SSQUECJMKSQDHG-UHFFFAOYSA-N
Synonym 4'-chloro-2'-fluoro-3'-methoxyacetophenone, 1-4-chloro-2-fluoro-3-methoxyphenyl ethanone
PubChem CID 66523544
IUPAC Name 1-(4-chloro-2-fluoro-3-methoxyphenyl)ethanone
SMILES COC1=C(Cl)C=CC(C(C)=O)=C1F
Specifications
Quantity 250 mg
Formula Weight 202.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4'-Chloro-2'-fluoro-3'-methoxyacetophenone