missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-3-methoxybenzamide, 97%, Thermo Scientific™

Catalog Number p-7032064
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1323966-12-6
C8H7ClFNO2
203.597
MFCD19687206
PGFYDZXJRFHQTG-UHFFFAOYSA-N
66523542
4-chloro-2-fluoro-3-methoxybenzamide
COC1=C(C=CC(=C1F)C(=O)N)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1323966-12-6
C8H7ClFNO2
203.597
MFCD19687206
PGFYDZXJRFHQTG-UHFFFAOYSA-N
66523542
4-chloro-2-fluoro-3-methoxybenzamide
COC1=C(C=CC(=C1F)C(=O)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-2-fluoro-3-methoxybenzamide
Quantità 250 mg
Peso formulazione 203.6
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'