missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorocinnamic acid, 97%, Thermo Scientific™

Catalog Number p-7032729
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1324063-26-4
C11H10ClFO3
244.646
MFCD19687219
PJYOROFBBRKJCT-GQCTYLIASA-N
4-chloro-3-ethoxy-2-fluorocinnamic acid, 2e-3-4-chloro-3-ethoxy-2-fluorophenyl prop-2-enoic acid, 3-4-chloro-3-ethoxy-2-fluorophenyl acrylic acid
66523597
(E)-3-(4-chloro-3-ethoxy-2-fluorophenyl)prop-2-enoic acid
CCOC1=C(C=CC(=C1F)C=CC(=O)O)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1324063-26-4
C11H10ClFO3
244.646
MFCD19687219
PJYOROFBBRKJCT-GQCTYLIASA-N
4-chloro-3-ethoxy-2-fluorocinnamic acid, 2e-3-4-chloro-3-ethoxy-2-fluorophenyl prop-2-enoic acid, 3-4-chloro-3-ethoxy-2-fluorophenyl acrylic acid
66523597
(E)-3-(4-chloro-3-ethoxy-2-fluorophenyl)prop-2-enoic acid
CCOC1=C(C=CC(=C1F)C=CC(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-3-ethoxy-2-fluorocinnamic acid
Quantità 250 mg
Peso formulazione 244.65
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'