Learn More
Alfa Aesar™ 4-Chloro-3-fluorocinnamic acid, 95%
Marca: Alfa Aesar™ H32636.09
Dettagli aggiuntivi : CAS : 202982-66-9 Peso : 0.01000kg
Quantità | 10g |
---|
Identificatori chimici
202982-66-9 | |
200.593 | |
MNELKRQRBRYHBV-DUXPYHPUSA-N | |
5708403 | |
C1=CC(=C(C=C1C=CC(=O)O)F)Cl |
C9H6ClFO2 | |
MFCD00143294 | |
4-chloro-3-fluorocinnamic acid, e-3-4-chloro-3-fluorophenyl acrylic acid, 2e-3-4-chloro-3-fluorophenyl prop-2-enoic acid, 3-4-chloro-3-fluoro-phenyl-acrylic acid, rarechem bk hw 0060, e-4-chloro-3-fluorocinnamic acid, 3-4-chloro-3-fluorophenyl acrylic acid, e-3-4-chloro-3-fluorophenyl prop-2-enoic acid | |
(E)-3-(4-chloro-3-fluorophenyl)prop-2-enoic acid |
Specifica
4-Chloro-3-fluorocinnamic acid | |
10g | |
C9H6ClFO2 | |
4-chloro-3-fluorocinnamic acid, e-3-4-chloro-3-fluorophenyl acrylic acid, 2e-3-4-chloro-3-fluorophenyl prop-2-enoic acid, 3-4-chloro-3-fluoro-phenyl-acrylic acid, rarechem bk hw 0060, e-4-chloro-3-fluorocinnamic acid, 3-4-chloro-3-fluorophenyl acrylic acid, e-3-4-chloro-3-fluorophenyl prop-2-enoic acid | |
C1=CC(=C(C=C1C=CC(=O)O)F)Cl | |
200.593 | |
200.6 |
241°C to 245°C | |
202982-66-9 | |
MFCD00143294 | |
MNELKRQRBRYHBV-DUXPYHPUSA-N | |
(E)-3-(4-chloro-3-fluorophenyl)prop-2-enoic acid | |
5708403 | |
95% |