missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-4'-methoxybutyrophenone, 97%, Thermo Scientific™

Catalog Number p-7022600
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
40877-19-8
C11H13ClO2
212.67
MFCD00018995
NGBTWDPPZFGUAY-UHFFFAOYSA-N
96414
4-chloro-1-(4-methoxyphenyl)butan-1-one
COC1=CC=C(C=C1)C(=O)CCCCl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
40877-19-8
C11H13ClO2
212.67
MFCD00018995
NGBTWDPPZFGUAY-UHFFFAOYSA-N
96414
4-chloro-1-(4-methoxyphenyl)butan-1-one
COC1=CC=C(C=C1)C(=O)CCCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-4'-methoxybutyrophenone
Punto di fusione 30°C to 33°C
Punti di ebollizione 159°C to 163°C (2 mmHg)
Punto d'infiammabilità >110°C (230°F)
Quantità 5 g
Beilstein 880054
Sinonimo 4-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl butan-1-one, acmc-20aml7, gamma-chloro-4'-methoxybutyrophenone, 4-chloro-1-4-methoxyphenyl-1-butanone, 1-butanone,4-chloro-1-4-methoxyphenyl, 4-chloro-1-4-methoxy-phenyl-butan-1-one, 1-butanone, 4-chloro-1-4-methoxyphenyl, 4-chloro-4 inverted exclamation marka-methoxybutyrophenone
Peso formulazione 212.68
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'