Learn More
4-Chloro-6-(2-methyl-1H-imidazol-1-yl)pyrimidine, TRC
Codice prodotto. 30364189
Cambia vista
Click to view available options
Quantità:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30364189 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30364189 Fornitore Toronto Research Chemicals N. del fornitore TRCC58754850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 4-Chloro-6-(2-methyl-1H-imidazol-1-yl)pyrimidine
CAS 941294-31-1
Formula molecolare C56 H84 O21 S2
Quantità 50 mg
InChI Formula InChI=1S/C56H84O21S2/c1-10-12-29(2)13-17-52(5,58)51-31(4)21-40-39(72-51)27-47-56(9,75-40)50(57)49-43(71-47)25-42-48(73-49)30(3)14-18-54(7)45(70-42)28-44-55(8,77-54)19-15-32-33(69-44)22-35-34(66-32)23-36-37(67-35)24-41-38(68-36)26-46(76-79(62,63)64)53(6,74-41)16-11-20-65-78(59,60)61/h10,13,17,30,32-51,57-58H,1-2,4,11-12,14-16,18-28H2,3,5-9H3,(H,59,60,61)(H,62,63,64)/b17-13+/t30?,32?,33?,34?,35?,36?,37?,38?,39?,40?,41?,42?,43?,44?,45?,46-,47?,48?,49?,50+,51?,52+,53+,54?,55?,56?/m0/s1
SMILES CC1CCC2(C)OC3(C)CCC4OC5CC6OC7C[C@H](OS(=O)(=O)O)[C@@](C)(CCCOS(=O)(=O)O)OC7CC6OC5CC4OC3CC2OC8CC9OC%10CC%11OC(C(=C)CC%11OC%10(C)[C@H](O)C9OC18)[C@](C)(O)\C=C\C(=C)CC=C
IUPAC Name [(13R,14S,34R)-34-hydroxy-40-[(2R,3E)-2-hydroxy-5-methylideneocta-3,7-dien-2-yl]-13,25,27,30,35-pentamethyl-39-methylidene-13-(3-sulfooxypropyl)-4,8,12,17,21,26,32,36,41,45,49-undecaoxaundecacyclo[25.22.0.03,25.05,22.07,20.09,18.011,16.031,48.033,46.035,44.037,42]nonatetracontan-14-yl] hydrogen sulfate
Molecular Weight (g/mol) 1157.384
Peso formulazione 1156.495