missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-phenylbutyramide, 97%, Thermo Scientific™

Catalog Number p-7040111
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7578-45-2
C10H12ClNO
197.662
MFCD01357318
GJMGKNWSRKDALN-UHFFFAOYSA-N
4-chloro-n-phenylbutyramide, n-phenyl-4-chlorobutanamide, 4-chloro-n-phenyl-butyramide
5106095
4-chloro-N-phenylbutanamide
C1=CC=C(C=C1)NC(=O)CCCCl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7578-45-2
C10H12ClNO
197.662
MFCD01357318
GJMGKNWSRKDALN-UHFFFAOYSA-N
4-chloro-n-phenylbutyramide, n-phenyl-4-chlorobutanamide, 4-chloro-n-phenyl-butyramide
5106095
4-chloro-N-phenylbutanamide
C1=CC=C(C=C1)NC(=O)CCCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Chloro-N-phenylbutyramide
Quantità 250 mg
Peso formulazione 197.66
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'