Learn More
4-Chloropyridine-2-carboxamide, TRC
Codice prodotto. 30350857
Cambia vista
Click to view available options
Quantità:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30350857 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30350857 Fornitore Toronto Research Chemicals N. del fornitore TRCC38147510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 4-Chloropyridine-2-carboxamide
CAS 99586-65-9
Formula molecolare C6H5ClN2O
Quantità 10 g
Sinonimo 4-Chloro-2-pyridinecarboxamide, 4-Chloropicolinamide, NSC 191958, 4-Chloro-pyridine-2-carboxylic Acid Amide
InChI Formula InChI=1S/C6H5ClN2O/c7-4-1-2-9-5(3-4)6(8)10/h1-3H,(H2,8,10)
SMILES NC(=O)c1cc(Cl)ccn1
IUPAC Name 4-chloropyridine-2-carboxamide
Molecular Weight (g/mol) 156.57
Peso formulazione 156.009