Learn More
(+/-)-4-Chlorostyrene oxide, 97+%
Product Code. 11475910
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475910 25 g 25g
11465910 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11475910 Supplier Thermo Scientific Alfa Aesar Supplier No. H26516.14
 Restricted Item
Spedizione stimata: 29-12-2026
Add to basket
CAS: 2788-86-5 | C8H7ClO | 154.593 g/mol
(±)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-4-Chlorostyrene oxide is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2788-86-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00956179
InChI Key IBWLXNDOMYKTAD-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl oxirane, 4-chlorostyrene oxide, p-chlorostyrene oxide, oxirane, 4-chlorophenyl, p-chlorophenyl oxirane, 2-4-chloro-phenyl-oxirane, +/--4-chlorostyrene oxide, benzene, 1-chloro-4-epoxyethyl, 4-chlorophenyl oxirane, 4-chloro styrene oxide
PubChem CID 197010
IUPAC Name 2-(4-chlorophenyl)oxirane
SMILES C1C(O1)C2=CC=C(C=C2)Cl
Specifications
Density 1.283
Quantity 25 g
Refractive Index 1.552
Boiling Point 230°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 154.59
Percent Purity 98%
Chemical Name or Material (+/-)-4-Chlorostyrene oxide