missing translation for 'onlineSavingsMsg'
Learn More

4'-Diethylaminoacetophenone, 99%

Codice prodotto. 11378046
Cambia vista
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11378046 5 g 5g
11368046 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11378046 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L13331.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5520-66-1 | C12H17NO | 191.27 g/mol

Used in synthesis of photocurable coatings and inks; coloring materials for recording media and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5520-66-1
Formula molecolare C12H17NO
Molecular Weight (g/mol) 191.27
Numero MDL MFCD00043651
InChI Key HMIBQFXWSUBFTG-UHFFFAOYSA-N
Sinonimo 4'-diethylaminoacetophenone, p-diethylaminoacetophenone, 4-diethylaminoacetophenone, 1-4-diethylamino phenyl ethanone, ethanone, 1-4-diethylamino phenyl, 1-acetyl-4-diethylamino benzene, acmc-1aspz, 4-acetyl-n,n-diethylaniline
PubChem CID 138529
IUPAC Name 1-[4-(diethylamino)phenyl]ethanone
SMILES CCN(CC)C1=CC=C(C=C1)C(C)=O

Specifications

Materiale o nome chimico 4'-Diethylaminoacetophenone
Punto di fusione 47°C to 49°C
Quantità 5 g
Beilstein 2093988
Peso formulazione 191.27
Percent Purity 99%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.