Learn More
4-(Dimethylamino)pyridine tribromide, 97%
Product Code. 15491118
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491118 5 g 5g
15401128 25 g 25g
15411128 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15491118 Supplier Thermo Scientific Alfa Aesar Supplier No. H55010.06
 Restricted Item
Spedizione stimata: 22-12-2026
Add to basket
CAS: 92976-81-3 | C7H10Br3N2- | 361.883 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 92976-81-3
Molecular Formula C7H10Br3N2-
Molecular Weight (g/mol) 361.883
MDL Number MFCD00043439
InChI Key TYDZHQMKSLAFKM-UHFFFAOYSA-N
Synonym 4-dimethylamino pyridine hydrobromide perbromide
PubChem CID 131863682
SMILES CN(C)C1=CC=NC=C1.Br[Br-]Br
Specifications
Melting Point 129°C to 132°C
Quantity 5 g
UN Number UN3261
Formula Weight 362.89
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)pyridine tribromide