Learn More
4'-Fluoro-2'-methoxyacetophenone, 97%
Product Code. 11392307
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392307 5 g 5g
11382307 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11392307 Supplier Thermo Scientific Alfa Aesar Supplier No. L09862.06
 Restricted Item
Spedizione stimata: 02-12-2026
Add to basket
CAS: 51788-80-8 | C9H9FO2 | 168.167 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51788-80-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00061143
InChI Key YOXBPWVWNQROBJ-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-methoxyphenyl ethanone, 4-fluoro-2-methoxyacetophenone, 4'-fluoro-2'-methoxyacetophenone, 1-4-fluoro-2-methoxyphenyl ethan-1-one, 2'-methoxy-4'-fluoroacetophenone, 1-4-fluoro-2-methoxy-phenyl-ethanone, 1-acetyl-4-fluoro-2-methoxybenzene, 2-methoxy-4-fluoroacetophenone, 4'-fluoro-2'-methoxyacetophenone,, pubchem13459
PubChem CID 142887
IUPAC Name 1-(4-fluoro-2-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)OC
Specifications
Melting Point 51°C to 53°C
Quantity 5 g
Beilstein 2251986
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 4'-Fluoro-2'-methoxyacetophenone