missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic acid hydrochloride, 99%, Thermo Scientific™

Catalog Number p-7040585
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42823-46-1
C8H10ClN3O2
215.637
MFCD00040587
YETFLAUJROGBMC-UHFFFAOYSA-N
4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
3084875
4-(diaminomethylideneamino)benzoic acid;hydrochloride
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
42823-46-1
C8H10ClN3O2
215.637
MFCD00040587
YETFLAUJROGBMC-UHFFFAOYSA-N
4-guanidinobenzoic acid hydrochloride, n-4-carboxyphenyl guanidine hydrochloride, 4-guanidinobenzoic acid hcl, 4-guanidino benzoic acid hydrochloride, 4-amidinoamino benzoic acid, chloride, pubchem14981, pubchem14993, acmc-1an3m, ksc494a0j, 4-guanidino-benzoic acid-hcl salt
3084875
4-(diaminomethylideneamino)benzoic acid;hydrochloride
C1=CC(=CC=C1C(=O)O)N=C(N)N.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Guanidinobenzoic acid hydrochloride
Punto di fusione 285°C (decomposition)
Formula lineare H2NC(=NH)NHC6H4CO2H·HCl
Quantità 1 g
Beilstein 3718032
Peso formulazione 215.64
Percent Purity 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'