Learn More
4-Hydroxy-2-methylthio-6-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™
Codice prodotto. 15430636
Cambia vista
Click to view available options
Quantità:
1 g
5 g
25 g
Dimensione della confezione:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15430636 25 g 25g
15410636 1 g 1g
15420636 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 15430636 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H53382.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 16097-62-4
Formula molecolare C6H5F3N2OS
Molecular Weight (g/mol) 210.174
Numero MDL MFCD00153191
InChI Key BRLPDRZKOMZVOY-UHFFFAOYSA-N
Sinonimo 2-methylthio-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methylthio-6-trifluoromethyl pyrimidine, 2-methylsulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-methylsulfanyl-6-trifluoromethyl-4-pyrimidinol, methylsulfanyltrifluoromethylpyrimidinol, 2-methylsulfanyl-6-trifluoromethyl-1h-pyrimidin-4-one, 4-hydroxy-2-methylthio-6-trifluoromethyl-pyrimidine, pubchem16996, 2-methylthio-4-trifluoromethyl-6-pyrimidinol, 2-methylthio-6-trifluoromethylpyrimidin-4-ol
PubChem CID 248062
IUPAC Name 2-methylsulfanyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CSC1=NC(=O)C=C(N1)C(F)(F)F
Specifica
Materiale o nome chimico 4-Hydroxy-2-methylthio-6-(trifluoromethyl)pyrimidine
Punto di fusione 176°C to 180°C
Quantità 25 g
Peso formulazione 210.17
Percent Purity 97%