Learn More
4-Hydroxy-TEMPO Benzoate, Free Radical, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-Hydroxy-TEMPO Benzoate, Free Radical |
| CAS | 3225-26-1 |
| Formula molecolare | C26 H40 N2 O7 S . H Cl |
| Quantità | 500 mg |
| Sinonimo | 2,2,6,6-Tetramethyl-4-(benzoyloxy)piperidine-1-oxyl, 2,2,6,6-Tetramethyl-4-benzoatepiperidine-1-oxy, 2,2,6,6-Tetramethyl-4-benzoyloxypiperidine N-Oxide, 2,2,6,6-Tetramethyl-4-benzoyloxypiperidine-1-oxy Radical, 2,2,6,6-Tetramethyl-4-hydroxypiperidin-1-oxyl Benzoate, 2,2,6,6-Tetramethyl-4-hydroxypiperidine-1-oxyl Benzoate, 2,2,6,6-Tetramethylpiperidin-1-oxyl-4-yl Benzoate, 4-(Benzoyloxy)-2,2,6,6-tetramethylpiperidin-1-yloxyl, 4-(Benzoyloxy)-2,2,6,6-tetramethylpiperidine-1-oxyl, 4-(Benzoyloxy)-2,2,6,6-tetramethylpiperidine-N-oxyl, 4-Hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl Benzoate, 4-Hydroxy-2,2,6,6-tetramethylpiperidinyl-1-oxyl Benzoate, Tempol Benzoate, 4-(Benzoyloxy)-2,2,6,6-tetramethyl-1-piperidinyloxy, 1-Oxyl-2,2,6,6-tetramethylpiperidin-4-yl Benzoate, 1-Oxyl-4-benzoyloxy-2,2,6,6-tetramethylpiperidine, 4-(Benzoyloxy)-2,2,6,6-tetramethyl-1-piperidinyloxy, 1-Oxyl-2,2,6,6-tetramethylpiperidin-4-yl Benzoate, 1-Oxyl-4-benzoyloxy-2,2,6,6-tetramethylpiperidine, 4-Hydroxy-2,2,6,6-tetramethyl-piperidinooxy Benzoate (Ester) |
| InChI Formula | InChI=1S/C26H40N2O7S.ClH/c1-6-7-15-12-18(28(3)13-15)24(32)27-19(14(2)29)23-26(21(31)20(30)25(34-23)36-5)22(35-26)16-8-10-17(33-4)11-9-16;/h8-11,14-15,18-23,25,29-31H,6-7,12-13H2,1-5H3,(H,27,32);1H/t14-,15-,18+,19-,20-,21-,22?,23-,25-,26+;/m1./s1 |
| SMILES | Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)N[C@H]([C@@H](C)O)[C@H]2O[C@H](SC)[C@H](O)[C@@H](O)[C@@]23OC3c4ccc(OC)cc4 |
| IUPAC Name | (2S,4R)-N-[(1R,2R)-1-[(3S,4R,5R,6R,8R)-4,5-dihydroxy-2-(4-methoxyphenyl)-6-methylsulfanyl-1,7-dioxaspiro[2.5]octan-8-yl]-2-hydroxypropyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 561.131 |
| Peso formulazione | 560.232 |