missing translation for 'onlineSavingsMsg'
Learn More

4-Iodoanisole, 98%, Thermo Scientific Chemicals

Catalog Number p-4509686
22.13 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
25 g
100 g
500 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
696-62-8
C7H7IO
234.04
SYSZENVIJHPFNL-UHFFFAOYSA-N
4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
69676
1-iodo-4-methoxybenzene
COC1=CC=C(C=C1)I
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
696-62-8
C7H7IO
234.04
SYSZENVIJHPFNL-UHFFFAOYSA-N
4-iodoanisole, p-iodoanisole, benzene, 1-iodo-4-methoxy, isoform, 4-methoxyiodobenzene, p-methoxyiodobenzene, anisole, p-iodo, 4-iodomethoxybenzene, p-iodophenyl methyl ether, 1-iodo-4-methoxy-benzene
69676
1-iodo-4-methoxybenzene
COC1=CC=C(C=C1)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-Iodoanisole
Punto di fusione 46.0°C to 51.0°C
Colore Brown to White
Punti di ebollizione 237.0°C (726.0 mmHg)
Punto d'infiammabilità >110°C
Spettro a infrarossi Authentic
Intervallo percentuale saggio 97.5% min. (GC)
Imballaggio Glass bottle
Formula lineare IC6H4OCH3
Quantità 25 g
Beilstein 06,208
Peso formulazione 234.04
Percent Purity 98%
Forma fisica Crystalline Powder and/or Chunks
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'