missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzene-1,3-dicarboxylic Acid, Mikromol™

Codice prodotto. 30050780
Cambia vista
Click to view available options
Quantità:
25 mg
100 mg
4 x 25 mg
Dimensione della confezione:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30050780 25 mg 25mg
30051387 100 mg 100mg
30459638 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050780 Supplier Mikromol Supplier No. MM3421.010025

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methoxybenzene-1,3-dicarboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 4-methoxybenzene-1,3-dicarboxylic acid
CAS 2206-43-1
Nomi componenti o analita 4-methoxybenzene-1,3-dicarboxylic acid
Shipping Condition Room Temperature
Formula molecolare C9 H8 O5
Certificazione/Conformità ISO 17025
Sinonimo Picotamide Monohydrate Imp. A (EP), Picotamide Imp. A (EP), 2-Methoxy-1,5-benzenedicarboxylic acid, 4-Methoxy-1,3-benzenedicarboxylic acid, 4-Methoxyisophthalic acid, NSC 20694, Picotamide Monohydrate Impurity A, Picotamide Impurity A
InChI Formula InChI=1S/C9H8O5/c1-14-7-3-2-5(8(10)11)4-6(7)9(12)13/h2-4H,1H3,(H,10,11)(H,12,13)
SMILES COc1ccc(cc1C(=O)O)C(=O)O
IUPAC Name 4-methoxybenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol) 196.16
Quantità 25 mg
Peso formulazione 196.0372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.