Learn More
4-Methoxybenzyl Chloride (Stabilized), TRC
Specifications
Specifications
| Materiale o nome chimico | 4-Methoxybenzyl Chloride |
| CAS | 824-94-2 |
| Formula molecolare | C8 H9 Cl O |
| Quantità | 5 g |
| Sinonimo | p-(Chloromethyl)-anisole, 1-(Chloromethyl)-4-(methyloxy)benzene, 1-(Chloromethyl)-4-methoxybenzene, 4-(Chloromethyl)-1-methoxybenzene, 4-(Chloromethyl)anisole, 4-(Chloromethyl)phenyl Methyl Ether, 4-Methoxybenzyl Chloride, 4-Methyloxybenzyl Chloride, NSC 172955, p-(Chloromethyl)(methoxy)benzene, p-(Chloromethyl)anisole, p-Anisyl Chloride, p-Methoxybenzyl Chloride, α-Chloro-4-methoxytoluene, α-Chloro-p-methoxytoluene, PMB-Cl |
| InChI Formula | InChI=1S/C8H9ClO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3 |
| SMILES | COc1ccc(CCl)cc1 |
| IUPAC Name | 1-(chloromethyl)-4-methoxybenzene |
| Molecular Weight (g/mol) | 156.61 |
| Peso formulazione | 156.0342 |