Learn More
4-Methoxyphenacyl chloride, 97%
Product Code. 10508912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10508912 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10508912 Supplier Thermo Scientific Acros Supplier No. 425260010
 Restricted Item
Spedizione stimata: 10-12-2026
Add to basket
CAS: 2196-99-8 | C9H9ClO2 | 184.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 2196-99-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 98°C to 100°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9ClO2
Linear Formula H3COC6H4COCH2Cl
MDL Number MFCD00216508
Quantity 1 g
Synonym 2-chloro-1-4-methoxyphenyl ethanone, 4-methoxyphenacyl chloride, 2-chloro-4'-methoxyacetophenone, 2-chloro-4'-methoxylphacetone, 4-methoxyphenacylchloride, 2-chloro-1-4-methoxy-phenyl-ethanone, 2-chloro-1-4-methoxyphenyl ethan-1-one, ethanone, 2-chloro-1-4-methoxyphenyl, 4-chloroacetyl anisole, 4'-methoxy-2-chloroacetophenone
Solubility Information Solubility in water: negligible
InChI Key MCRINSAETDOKDE-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C(=O)CCl
IUPAC Name 2-chloro-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol) 184.62
PubChem CID 237806
Formula Weight 184.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenacyl chloride, 96.5%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxyphenacyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.