Learn More
4-Methyl-2-pentyne, 97%
Product Code. 15440258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440258 1 g 1g
15450258 5 g 5g
15460258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15440258 Supplier Thermo Scientific Alfa Aesar Supplier No. H53442.03
 Restricted Item
In stock
Add to basket
CAS: 21020-27-9 | C6H10 | 82.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21020-27-9
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00041615
InChI Key SLMFWJQZLPEDDU-UHFFFAOYSA-N
Synonym 4-methyl-2-pentyne, 2-pentyne, 4-methyl, isopropylmethylacetylene, acmc-1co71
PubChem CID 140789
IUPAC Name 4-methylpent-2-yne
SMILES CC#CC(C)C
Specifications
Density 0.711
Boiling Point 71°C to 73°C
Flash Point −12°C (−10°F)
Linear Formula (CH3)2CHC≡CCH3
Refractive Index 1.407
Quantity 1 g
UN Number UN3295
Formula Weight 82.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-pentyne
compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-pentyne
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methyl-2-pentyne