Learn More
4-(methylsulfanyl)butanal, TRC
Codice prodotto. 30478719
Cambia vista
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30478719 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30478719 Fornitore Toronto Research Chemicals N. del fornitore TRCM10604510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 4-(methylsulfanyl)butanal
CAS 42919-64-2
Formula molecolare C5H10OS
Quantità 10 mg
InChI Formula InChI=1S/C5H10OS/c1-7-5-3-2-4-6/h4H,2-3,5H2,1H3
SMILES CSCCCC=O
IUPAC Name 4-methylsulfanylbutanal
Molecular Weight (g/mol) 118.19
Peso formulazione 118.0452