Learn More
MedChemExpress 4-Methylumbelliferyl-β-D-glucuronide hydrate
Specifica
Specifica
| CAS | 881005-91-0 |
| Solubilità | DMSO : 150 mg/mL (405.07 mM∣Need ultrasonic) ∣H2O : 5.2 mg/mL (14.04 mM∣Need ultrasonic and warming) |
| Sinonimo | MUG |
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Da utilizzare con (applicazione) | Neuroscience-Neuromodulation |
| Quantità | 25 mg |
| Peso molecolare | 370.31 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 99.93% |
| Shipping Condition | Room temperature |