Learn More
4-Nitrobenzenemethanol, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-Nitrobenzenemethanol |
| CAS | 619-73-8 |
| Formula molecolare | C7H7NO3 |
| Quantità | 10 g |
| Sinonimo | (4-Nitrophenyl)methanol, 1-(Hydroxymethyl)-4-nitrobenzene, 4-(Hydroxymethyl)nitrobenzene, 4-Nitrobenzenemethanol, 4-Nitrobenzyl alcohol, NSC 5389, p-(Hydroxymethyl)nitrobenzene, p-Nitrobenzyl alcohol, p-Nitrophenylmethanol, p-Nitrobenzyl Alcohol |
| InChI Formula | InChI=1S/C7H7NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4,9H,5H2 |
| SMILES | OCc1ccc(cc1)[N+](=O)[O-] |
| IUPAC Name | (4-nitrophenyl)methanol |
| Molecular Weight (g/mol) | 153.14 |
| Peso formulazione | 153.0426 |