missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzonitrile, 97%, Thermo Scientific™

Catalog Number 15447537
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
401892-85-1
C7H4F5NS
229.168
MFCD03788521
SGACKKUEROEDNX-UHFFFAOYSA-N
4-pentafluorosulfanyl benzonitrile, 4-pentafluorothio benzonitrile, 4-cyanophenylsulphur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl benzonitrile, 4-pentafluoro-lambda∼6∼-sulfanyl benzonitrile, 4-pentafluoro-??-sulfanyl benzonitrile, 4-pentafluorosulfanyl cyanobenzene, 4-pentafluorosulphanyl benzonitrile, 4-pentafluoro-sulfanyl benzonitrile, 4-pentakis fluoranyl-$l^ 6-sulfanyl benzenecarbonitrile
2779212
4-(pentafluoro-$l^{6}-sulfanyl)benzonitrile
C1=CC(=CC=C1C#N)S(F)(F)(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
401892-85-1
C7H4F5NS
229.168
MFCD03788521
SGACKKUEROEDNX-UHFFFAOYSA-N
4-pentafluorosulfanyl benzonitrile, 4-pentafluorothio benzonitrile, 4-cyanophenylsulphur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl benzonitrile, 4-pentafluoro-lambda∼6∼-sulfanyl benzonitrile, 4-pentafluoro-??-sulfanyl benzonitrile, 4-pentafluorosulfanyl cyanobenzene, 4-pentafluorosulphanyl benzonitrile, 4-pentafluoro-sulfanyl benzonitrile, 4-pentakis fluoranyl-$l^ 6-sulfanyl benzenecarbonitrile
2779212
4-(pentafluoro-$l^{6}-sulfanyl)benzonitrile
C1=CC(=CC=C1C#N)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 4-(Pentafluorothio)benzonitrile
Quantità 5 g
Numero UN UN3439
Peso formulazione 229.17
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'