Descrizione
- Compatible with CentriVap Pro Vacuum Concentrators
- Stainless steel material
- Optional PTFE coated
Specifica
Specifica
| Materiale | Stainless Steel |
| Tipo | Rotor |
| Material (Coating) | PTFE |
| Sistema rotore | CentriVap Pro |
| Dimensioni (L x P x A) | 25.4 x 15.75 x 7.62 cm (10 x 6.2 x 3 in.) |