Learn More
4-(Trifluoromethyl)benzonitrile, TRC
Specifica
Specifica
| Materiale o nome chimico | 4-(Trifluoromethyl)benzonitrile |
| CAS | 455-18-5 |
| Formula molecolare | C8H4F3N |
| Quantità | 10 g |
| Sinonimo | 4-(Trifluoromethyl)benzonitrile, 4-Cyano-α,α,α-trifluorotoluene, 4-Trifluoromethyl-1-cyanobenzene, NSC 88340, p-(Trifluoromethyl)benzonitrile, p-(Trifluoromethyl)cyanobenzene, p-Cyanobenzotrifluoride, α,α,α-Trifluoro-p-tolunitrile |
| InChI Formula | InChI=1S/C8H4F3N/c9-8(10,11)7-3-1-6(5-12)2-4-7/h1-4H |
| SMILES | FC(F)(F)c1ccc(cc1)C#N |
| IUPAC Name | 4-(trifluoromethyl)benzonitrile |
| Molecular Weight (g/mol) | 171.12 |
| Peso formulazione | 171.0296 |