Learn More
Labcon™ SuperClear™ 5 mL Centrifuge Tubes with Attached Caps
Product Code. 16674012 Shop All Labcon Products
Change view
Click to view available options
Color:
Amber
Assorted
Natural
Unit Size:
Pack of 200
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16674012 Natural Pack of 250
16684012 Assorted Pack of 200
17567896 Amber Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16674012 Supplier Labcon™ Supplier No. 30488600009
  • ONLINE EXCLUSIVE PRICE
    49.25€
    41.91€ / pezzi 250
    Risparmia 7.34€
    14% Sconto
In stock
Add to basket
Edge
Add to basket
Centrifuge tubes with attached flat snap caps, molded graduations, and writing surface.
  • Freezable to -80°C
  • Made in USA
Specifications
Cap Style Attached
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Autoclavable Autoclavable
Capacity (Metric) 5 mL
Color Natural
Material Polypropylene Resin
For Use With (Application) Centrifugation
Packaging 250 Tubes/Bag, 10 Bags/Cs.
Clean Claims ATP, RNase/DNase and Endotoxin-free
Sterility Non-sterile
Product Line SuperClear
Graduated Yes
Bottom Shape Conical
Max. RCF 25,000 RCF