missing translation for 'onlineSavingsMsg'
Learn More

5(6)-Tetramethylrhodamine isothiocyanate

Codice prodotto. 15494659
Cambia vista
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15494659 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15494659 Fornitore Thermo Scientific Alfa Aesar N. del fornitore J62258.MA
Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 95197-95-8 | C25H21N3O3S

5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-based applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-bas
TRUSTED_SUSTAINABILITY

Specifica

Materiale o nome chimico 5(6)-Tetramethylrhodamine isothiocyanate
CAS 95197-95-8
Colore Purple to Red
Formula molecolare C25H21N3O3S
Numero MDL MFCD00063517
Quantità 10 mg
Sensibilità Light, Air and Moisture Sensitive
Sinonimo TRITC; MRITC
Informazioni di solubilità Soluble in DMSO or methanol
Peso formulazione 443.52
Forma fisica Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.