missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-1,6-naphthyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-4229168
290.01 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 g
5 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
337958-60-8
C8H4Cl2N2
199.04
HNRPBMNTCYRAJD-UHFFFAOYSA-N
pubchem14653, 5,7-dichloro-1,6-napthyridine, 1,6-naphthyridine,5,7-dichloro, 1,6-naphthyridine, 5,7-dichloro, 5,7-bis chloranyl-1,6-naphthyridine
12204233
5,7-dichloro-1,6-naphthyridine
C1=CC2=C(N=C(C=C2N=C1)Cl)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
337958-60-8
C8H4Cl2N2
199.04
HNRPBMNTCYRAJD-UHFFFAOYSA-N
pubchem14653, 5,7-dichloro-1,6-napthyridine, 1,6-naphthyridine,5,7-dichloro, 1,6-naphthyridine, 5,7-dichloro, 5,7-bis chloranyl-1,6-naphthyridine
12204233
5,7-dichloro-1,6-naphthyridine
C1=CC2=C(N=C(C=C2N=C1)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 5, 7-Dichloro-1, 6-naphthyridine
CAS min. % 96.0
CAS max % 100.0
Punto di fusione 138°C to 140°C
Colore White to Yellow
Spettro a infrarossi Authentic
Intervallo percentuale saggio 96% min. (GC)
Imballaggio Glass bottle
Quantità 1 g
Peso formulazione 199.04
Percent Purity 97%
Forma fisica Crystalline Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'