Learn More
5-Acetamidonaphthalene-1-sulfonyl Chloride, TRC
Codice prodotto. 30327430
Cambia vista
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30327430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30327430 Fornitore Toronto Research Chemicals N. del fornitore TRCA1584651G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 5-Acetamidonaphthalene-1-sulfonyl Chloride
CAS 52218-37-8
Formula molecolare C12 H10 Cl N O3 S
Quantità 1 g
InChI Formula InChI=1S/C12H10ClNO3S/c1-8(15)14-11-6-2-5-10-9(11)4-3-7-12(10)18(13,16)17/h2-7H,1H3,(H,14,15)
SMILES CC(=O)Nc1cccc2c(cccc12)S(=O)(=O)Cl
IUPAC Name 5-acetamidonaphthalene-1-sulfonyl chloride
Molecular Weight (g/mol) 283.73
Peso formulazione 283.007