Learn More
5-Amino-1,2,3-thiadiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1,2,3-thiadiazole |
| CAS | 4100-41-8 |
| Molecular Formula | C2 H3 N3 S |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C2H3N3S/c3-2-1-4-5-6-2/h1H,3H2 |
| SMILES | Nc1cnns1 |
| IUPAC Name | thiadiazol-5-amine |
| Molecular Weight (g/mol) | 101.13 |
| Formula Weight | 101.0048 |