Learn More
5-Cholesten-24(RS)-methyl-d3-3beta-ol, CDN
Product Code. 30316655
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316655 0.01 g 0.01g
30315266 0.005 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316655 Supplier CDN Supplier No. CDND63720.01G
 Restricted Item
Spedizione stimata: 03-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Campesterol-d3 (Mixture of Diastereomers)
Molecular Formula C28 D3 H45 O
Quantity 0.01 g
Synonym (24ξ)-24-(Methyl-d3)cholest-5-en-3β-ol, (3β,24ξ)-Ergost-5-en-3-ol-D3 (24-methyl-D3), 5-Cholesten-24(RS)-methyl-d3-3β-ol
InChI Formula InChI=1 S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29 H,7-8,10-17H2,1-6H3/t19?,20-,22+,23+,24-,25+,26+,27+,28-/m1/s1/i3D3
SMILES [2 H]C([2 H])([2 H])C(CC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C(C)C
IUPAC Name (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6-methyl-5-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 403.6986
Formula Weight 403.3893 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity