missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-2-nitrobenzotrifluoride, 98+%

Marca:  Alfa Aesar™ A18780.14

Dettagli aggiuntivi : CAS : 393-09-9 Peso : 0.02500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantità 25g

 vedi altre versioni di questo prodotto

Codice prodotto. 11403068

  • 125.00€ / 25g


In stock, pronto per la spedizione: 1
Aggiungi al carrello

Descrizione

Descrizione

Applications
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifica

Identificatori chimici

393-09-9
209.1
WMQOSURXFLBTPC-UHFFFAOYSA-N
223094
C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]
C7H3F4NO2
MFCD00039741
5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
4-fluoro-1-nitro-2-(trifluoromethyl)benzene

Specifica

5-Fluoro-2-nitrobenzotrifluoride
88°C (190°F)
87°C to 88°C (2mmHg)
25g
C7H3F4NO2
UN2811
5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
WMQOSURXFLBTPC-UHFFFAOYSA-N
4-fluoro-1-nitro-2-(trifluoromethyl)benzene
223094
≥98%
1.497
1.46
22°C to 24°C
393-09-9
MFCD00039741
3304470
Soluble in chloroform and methanol.
C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]
209.1
209.1
Videos
SDS Documenti