missing translation for 'onlineSavingsMsg'
Learn More

5-Formylfuran-2-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7047187
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
27329-70-0
C5H5BO4
139.901
MFCD01114696
JUWYQISLQJRRNT-UHFFFAOYSA-N
2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid, 5-formyl-2-furanylboronic acid, 2-formylfuran-5-boronicacid, 5-formyl-2-furyl boronic acid, 5-formyl-2-furanyl boronic acid, boronic acid, 5-formyl-2-furanyl
2734355
(5-formylfuran-2-yl)boronic acid
B(C1=CC=C(O1)C=O)(O)O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
27329-70-0
C5H5BO4
139.901
MFCD01114696
JUWYQISLQJRRNT-UHFFFAOYSA-N
2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid, 5-formyl-2-furanylboronic acid, 2-formylfuran-5-boronicacid, 5-formyl-2-furyl boronic acid, 5-formyl-2-furanyl boronic acid, boronic acid, 5-formyl-2-furanyl
2734355
(5-formylfuran-2-yl)boronic acid
B(C1=CC=C(O1)C=O)(O)O

As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 5-Formylfuran-2-boronic acid
Punto di fusione ∽140°C (decomposition)
Quantità 1 g
Beilstein 1637050
Sensibilità Air Sensitive
Informazioni di solubilità Insoluble in water.
Peso formulazione 139.9
Percent Purity 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'