Learn More
5-Methyl-bis-(2-aminophenoxymethylene)-N,N,N’,N’-tetraacetate Methyl Ester, TRC
Codice prodotto. 30469093
Cambia vista
Click to view available options
Quantità:
50 mg
Dimensione della confezione:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30469093 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30469093 Fornitore Toronto Research Chemicals N. del fornitore TRCM29414050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 5-Methyl-bis-(2-aminophenoxymethylene)-N,N,N',N'-tetraacetate Methyl Ester
CAS 96315-10-5
Formula molecolare C27H34N2O10
Quantità 50 mg
Sinonimo 5-Methyl-Bis-(2-Aminophenoxymethylene)-N,N,N,N-Tetraacetate Methyl Ester
InChI Formula InChI=1S/C27H34N2O10/c1-19-10-11-21(29(17-26(32)36-4)18-27(33)37-5)23(14-19)39-13-12-38-22-9-7-6-8-20(22)28(15-24(30)34-2)16-25(31)35-3/h6-11,14H,12-13,15-18H2,1-5H3
SMILES COC(=O)CN(CC(=O)OC)c1ccccc1OCCOc2cc(C)ccc2N(CC(=O)OC)CC(=O)OC
IUPAC Name methyl 2-[2-[2-[2-[bis(2-methoxy-2-oxoethyl)amino]-5-methylphenoxy]ethoxy]-N-(2-methoxy-2-oxoethyl)anilino]acetate
Molecular Weight (g/mol) 546.57
Peso formulazione 546.2213